EMAIL THIS PAGE TO A FRIEND
X0751 Sigma

L-Xylulose

~95%, syrup

Synonym: threo-2-Pentulose, L-threo-Pentulose

  • CAS Number 527-50-4

  • Empirical Formula (Hill Notation) C5H10O5

  • Molecular Weight 150.13

  •  MDL number MFCD00075347

  •  PubChem Substance ID 24902124

Purchase

Properties

Related Categories Carbohydrate, Cell Biology, Metabolic Pathways, Metabolites and Cofactors on the Metabolic Pathways Chart, Metabolomics More...
InChI Key   ZAQJHHRNXZUBTE-UCORVYFPSA-N
assay   ~95%
form   syrup
color   light yellow
storage temp.   2-8°C

Description

Biochem/physiol Actions

L-Xylulose, ketopentose monosaccharide, is a nonpshosphorylated intermediate in the glucoronic acid pathway. L-Xylulose accumulates in the urine patients with pentosuria and cirrhosis.

L-xylulose is a monosaccharide which contains five carbon atoms, and includes a ketone functional group.

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
3
Flash Point(F) 
235.4 °F
Flash Point(C) 
113 °C

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?