• USA Home
  • X1378 - β-Xylosidase from Escherichia coli

EMAIL THIS PAGE TO A FRIEND
X1378 Sigma

β-Xylosidase from Escherichia coli

recombinant, expressed in E. coli, buffered aqueous solution

Synonym: 1,4-β-D-Xylanxylohydrolase, Exo-1,4-β-D-xylosidase

Purchase

Properties

Related Categories 3.2.x.x Glycosidases, 3.x.x.x Hydrolases, Application Index, Biochemicals and Reagents, Carbohydrate Hydrolysis,
recombinant   expressed in E. coli
form   buffered aqueous solution
foreign activity   β-galactosidase, α-mannosidase, β-hexosaminidase, α-fucosidase, neuraminidase, and proteases, none detected
storage temp.   2-8°C

Description

Unit Definition

One unit will hydrolyze 1.0 μmole of p-nitrophenyl β-D-xyloside to p-nitrophenol and D-xylose per min at pH 5.0 at 37 °C.

Physical form

Solution in 20 mM Tris HCl, pH 7.5, 25 mM NaCl.

Price and Availability


Amplified Detection

Biomedical Applications
Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles

Related Content

Enzymes & Proteins

Application Index | Enzyme Index | Substrate Index | Inhibitor Index | Cofactor Index | Lectin Index
Keywords: Cell signaling, Diagnostic, Drug discovery, Molecular biology

Peer-Reviewed Papers
15

References

Related Products

substrate

Product #

Description

Add to Cart

M7008 4-Methylumbelliferyl-β-D-xylopyranoside, β-xylosidase substrate

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?