• USA Home
  • X3750 - D-Xylulose 5-phosphate monosodium salt

EMAIL THIS PAGE TO A FRIEND
X3750 Sigma

D-Xylulose 5-phosphate monosodium salt

~80% (enzymatic)

Synonym: Xu-5-P monosodium salt

Purchase

Properties

InChI Key   OVHWWORCLJVDMI-TYSVMGFPSA-M
assay   ~80% (enzymatic)
storage temp.   −20°C

Description

Biochem/physiol Actions

Xylulose-5-phoshate (Xu-5-P) is a metabolite of glucose via the pentose phosphate pathway. In the presence of excess glucose, liver Xu-5-P may act as a signaling molecule that regulates the nuclear transport of carbohydrate response element binding protein (ChREBP), a transcription factor that induces the expression of enzymes involved in glucose metabolism and lipogenesis.

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?