EMAIL THIS PAGE TO A FRIEND
XH Sigma

Tergitol

Type XH

Purchase

Properties

type   Type XH

Description

Other Notes

Polyglycol ether (nonionic) surfactants.

Legal Information

TERGITOL is a trademark of The Dow Chemical Company ("Dow") or an affiliated company of Dow

Price and Availability

Safety & Documentation

Safety Information

Personal Protective Equipment 
WGK Germany 
1
RTECS 
MD0911000
Flash Point(F) 
235.4 °F
Flash Point(C) 
113 °C

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?