XP725 Sigma

Panorama® Antibody Microarray - XPRESS Profiler725 Kit

DOWNLOAD MSDS (PDF)

Properties

Related Categories Antibody Arrays, Cell Biology, General Stem Cell Biology, Glial Markers, Neural Stem Cell Antibodies and Tracking,
species reactivity   human, mouse, rat
shipped in   wet ice
storage temp.   2-8°C

Description

Components

725 antibodies spotted in 32 sub-arrays, each containing duplicate spots of 23 antibodies, as well as duplicate positive control spots (a monoclonal antibody that recognizes Cy3 and Cy5) and several negative controls. Antibodies are spotted in high density to ensure strong signals. Antibodies are also available individually from Sigma. Refer to Antibody List for specific catalog number(s).

Application

The array can be used for comparing protein expression profile between two samples (test vs. reference samples). Each sample is directly labeled with a different florescent dye and the two samples are applied simultaneously on the array. The expression profile of each sample is recorded individually and compared. This allows one to determine if proteins are up- or down-regulated, comparatively, or allows for the profile of a sample to determine if a protein is present or not, such as when looking at genomic modifications.

Compatibility

Cell or tissue extracts, blood plasma or serum

General description

The Panorama® Antibody Microarray – XPRESS Profiler725 Kit is a proteomics tool that measures the relative level of protein expression (protein microarrays) in cell or tissue extracts and blood plasma or serum. The antibodies used represent families of proteins known to be involved in a variety of different biological pathways and molecular processes, including but not limited to cell stress, cell cycle, signaling transduction, apoptosis, neurobiology, and gene regulation. Results obtained using the array are qualitative and should be further evaluated by other methods such as immunoblotting assays or ELISA.

Other Notes

Learn more about the XPRESS Profiler725 array at www.sigma.com/xp725. For expression studies involving custom peptide libraries, visit www.sigma-aldrich.com/pepscreen.

Please note that featured antibody content is lot specific and is subject to change.

To view Panorama® array literature, visit www.sigma-aldrich.com/arrayliterature

Physical form

Nitrocellulose glass slide, proprietary treatment to minimize background staining

Features and Benefits

• An efficient and sensitive method of high throughput protein expression analysis
• Global solution to understanding protein relationships
• All antibodies are available separately for more extensive analysis of your samples

Legal Information

Panorama is a registered trademark of Sigma-Aldrich Co. LLC

Price and Availability

Kit component also available separately

Description

Product #

Add to Cart

Extraction/Labeling Buffer 30 ML E0655
Phosphatase Inhibitor Cocktail 2, aqueous solution (dark coloration may develop upon storage, which does not affect the activity) 0.3 ML P5726
Phosphate buffered saline, pH 7.4, contains TWEEN® 20, dry powder 1 EA P3563

Kit component only

Description

Product #

Add to Cart

Panorama® Antibody Slides - XPRESS Profiler725 2 EA    
QuadriPERM® Cell Culture Vessel , storage temp.: 2-8°C 2 EA    
Protease Inhibitor Cocktail 1 VL    
Benzonase®, Ultrapure 1000 UN    
Array Incubation Buffer 20 ML    
SigmaSpin Post-Reaction Clean-Up Columns 8 EA    
Microcentrifuge collection tube, size 2 mL 16 EA    
Panorama® Antibody List - XPRESS Profiler725 1 EA    

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?