EMAIL THIS PAGE TO A FRIEND
Y0500 Sigma

Select Yeast Extract

BioReagent, suitable for cell culture, powder

Purchase

Properties

Related Categories Cell Culture, Extracts, Media Components, Microbial Media for Molecular Biology and Protein Expression, Miscellaneous Reagents and Supplements,
product line   BioReagent
form   powder
solubility   H2O: 10 mg/mL hazy
suitability   suitable for cell culture

Description

Other Notes

Spray dried, autolyzed yeast extract

Water soluble portion of autolyzed yeast with intact B-complex vitamins. Yeast extract is a mixture of amino acids, peptides, water soluble vitamins and carbohydrates and can be used as additive for culture media.

Quality

May contain black specks.

Application

Yeast extract is used as a bacteriological media component for a variety of microorganisms and molecular genetics applications.

Price and Availability


Laboratory Gloves

All labs need water
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
RTECS 
ZF6610000

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?