• USA Home
  • Y1001 - Yeast Extract Hy-Yest® 412

EMAIL THIS PAGE TO A FRIEND
Y1001 Sigma

Yeast Extract Hy-Yest® 412

Hy-Yest® 412

Synonym: Select Yeast Extract

Purchase

Properties

Related Categories Biochemicals and Reagents, Protein Extracts, Proteins and Derivatives More...
form   powder
nitrogen analysis   ~5.2% amino, ~10.7% total

Description

Application

Recommended for use when low salt conditions are required.

Yeast extract is used as a bacteriological media component for a variety of microorganisms and molecular genetics applications.

Other Notes

Water soluble portion of autolyzed yeast with intact B-complex vitamins. Yeast extract is a mixture of amino acids, peptides, water soluble vitamins and carbohydrates and can be used as additive for culture media.

Legal Information

Hy-Yest is a registered trademark of Kerry Group

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?