EMAIL THIS PAGE TO A FRIEND
Y3500 Sigma

Yttrium(III) oxide

Purchase

Properties

Related Categories Materials Science, Metal and Ceramic Science, Oxides, Yttrium

Price and Availability


Laboratory Gloves
Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
1
RTECS 
ZG3850000

Documents

Certificate of Analysis

Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

204927 Yttrium(III) oxide, 99.999% trace metals basis

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?