• USA Home
  • Y4377 - YEEI Peptide, Phosphorylated trifluoroacetate salt

EMAIL THIS PAGE TO A FRIEND
Y4377 Sigma

YEEI Peptide, Phosphorylated trifluoroacetate salt

lyophilized powder

Synonym: (pY)EEI, YEEI Peptide

Purchase

Properties

assay   ≥95% (HPLC)
form   lyophilized powder
mol wt   calculated mol wt 1474.8 Da (free base)
  calculated mol wt 1588.8 Da
solubility   H2O: soluble
storage temp.   −20°C

Description

Amino Acid Sequence

Glu-Pro-Gln-Tyr(PO3H2)-Glu-Glu-Ile-Pro-Ile-Tyr-Leu

Biochem/physiol Actions

Used to study the effect of activation of endogenous c-Src tyrosine kinase.

YEEI Peptide is a phosphopeptide that activates Src family members by binding to their SH2 domain with high affinity.

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?