• USA Home
  • Z0134 - Anti-Zip Kinase antibody produced in rabbit

EMAIL THIS PAGE TO A FRIEND
Z0134 Sigma

Anti-Zip Kinase antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Properties

Related Categories Alphabetical Index, Antibodies, Antibodies for Apoptosis, Antibodies for Cell Biology, Antibodies for Kinase/Phosphatase Biology,
species reactivity   human
application(s)   microarray: suitable
  western blot: 0.5 μg/mL using human HeLa cell extract
clone   polyclonal
antibody form   affinity isolated antibody
form   buffered aqueous solution
mol wt   antigen mol wt 52 kDa
shipped in   dry ice
storage temp.   −20°C
Gene Information   human ... DAPK3(1613)
biological source   rabbit
conjugate   unconjugated

Description

Immunogen

synthetic peptide corresponding to amino acids 279-298 of an internal domain of human ZIP kinase

General description

Zipper-interacting protein kinase (also known as Dlk) is almost 81% homologous to death-associated protein kinase (DAP). Overexpression of this serine-threonine kinase induces apoptosis. Homodimerization of leucine zipper at the C-terminal leads to the activation of ZIP kinase. Activated ZIP kinase interacts with ATF-4, an oxidative stress-inducible transcription factor. ZIP kinase is also reported to interact with Par-4, a protein that modulates WT1 and results in reorganization of actin filaments and morphological changes associated with apoptosis. Anti-ZIP kinase specifically recognizes human ZIP kinase (52 kDa).

Physical form

Solution in phosphate buffered saline, containing 0.02% sodium azide.

Specificity

The antibody recognizes ZIP kinase, a serine/threonine kinase which possesses a leucine zipper that mediates interaction with transcription factor ATF-4, hence its name. Due to its homology to DAP kinase, it was also named DAP like kinase, Dlk.

Application

Anti-ZIP kinase may be used in immunoblotting applications at a working dilution of 1:1000 or 0.5 μg/mL using HeLa whole cell lysates. The antibody is also suitable for protein microarray.

Features and Benefits

Antibody Bioguarantee
Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Price and Availability


Antibody Explorer

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?