• USA Home
  • Z0751 - Zeatin riboside mixed Isomers

EMAIL THIS PAGE TO A FRIEND
Z0751 Sigma

Zeatin riboside mixed Isomers

~95%

  • CAS Number 28542-78-1

  • Empirical Formula (Hill Notation) C15H21N5O5

  • Molecular Weight 351.36

  •  MDL number MFCD00036809

  •  PubChem Substance ID 24902169

Purchase

Properties

Related Categories Biochemicals and Reagents, Nucleoside Analogs, Nucleosides, Nucleotides, Oligonucleotides More...
InChI Key   GOSWTRUMMSCNCW-HNNGNKQASA-N
assay   ~95%
storage temp.   −20°C

Description

Other Notes

Consists of an approximately equal mixture of cis- and trans-isomers; actual percentage given on the label.

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?