• USA Home
  • Z227498 - Millex® syringe filter units, disposable, Durapore® PVDF

EMAIL THIS PAGE TO A FRIEND
Z227498 Sigma

Millex® syringe filter units, disposable, Durapore® PVDF

pore size 0.22 μm, diam. 13 mm, non-sterile

Purchase

Properties

Related Categories Filtration Supplies, Labware, Manufacturer Browser, Millex syringe filter units, disposable, Durapore PVDF, Millipore,
material   PVDF membrane
  polypropylene housing
sterility   non-sterile
mfr. no.   Millipore SLGVX13NL
packaging   pack of 100 ea
diam.   13 mm
volume   ≤10 mL
pore size   0.22 μm
fitting   inlet female Luer (lock)
  outlet male Luer

Description

Application

Protein solutions; aqueous HPLC samples

Ultra-low protein binding (<1 μg/cm2). Component materials meet USP XXI Class VI Biological Tests for plastics. Water extractables <0.5%

General description

Units contain Durapore Hydrophilic PVDF filter membranes in plastic housings with Luer lock inlet and Luer slip outlet fittings.

Units smaller than 25 mm diameter are specifically designed to minimize hold-up in filtration of small volumes. The 3-4 mm units are recommended for volumes of <1 mL; the 13 mm units are recommended for volumes of 0.5 - 10 mL.

Legal Information

Durapore is a registered trademark of Merck KGaA, Darmstadt, Germany

Millex is a registered trademark of Merck KGaA, Darmstadt, Germany

Price and Availability


Empower your Cancer Research
Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis

Protocols & Articles

Articles

HPLC Analysis of Yohimbe Extract Adulterated with Erectile Dysfunction Drugs on Ascentis® Express RP-Amide

From our library of Articles, Sigma-Aldrich presents HPLC Analysis of Yohimbe Extract Adulterated with Erectile Dysfunction Drugs on Ascentis® Express RP-Amide
Keywords: Chromatography, Filtration, High performance liquid chromatography, Sample preparations, Vitamins

Sterile Millex® Filters for Cell Culture

Millex syringe filters are ideal for sterilizing 1 mL – 200 mL of cell culture reagents, such as cell culture media, antibiotics, supplements, sera, viruses for infection, and other biological soluti...
Keywords: Antibiotics, Capillary electrophoresis, Cell culture, Culture media, Filtration, Growth factors, Pharmaceutical, Sterilizations

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?