• USA Home
  • Z227528 - Millex® syringe filter units, disposable, PTFE
EMAIL THIS PAGE TO A FRIEND
Z227528 Sigma
Millex® syringe filter units, disposable, PTFE
pore size 0.20 μm, diam. 25 mm, Millipore SLFG025NS, Fluoropore PTFE (F) membrane
Purchase
Properties
Related Categories Filtration Supplies, Labware, Manufacturer Browser, Millex syringe filter units, disposable, PTFE, Millipore,
material   Fluoropore PTFE (F) membrane
  PTFE membrane
  high-density polyethylene housing
sterility   non-sterile
packaging   pack of 50 ea
mfr. no.   Millipore SLFG025NS
diam.   25 mm
volume   ≤100 mL
pore size   0.20 μm
fitting   inlet female Luer (lock)
  outlet male Luer
Description
Application
HPLC, for organic solvents and solutions
General description
All units contain PTFE filter membranes in plastic housings with Luer lock inlet and Luer slip outlet fittings.
Units smaller than 25 mm diameter are specifically designed to minimize hold-up in filtration of small volumes. The 3-4 mm units are recommended for volumes of <1 mL; the 13 mm units are recommended for volumes of 0.5-10 mL.
Suitability
Remove fine particles from organic solutions. Also use for venting applications.
Legal Information
Millex is a registered trademark of Merck KGaA, Darmstadt, Germany
Price and Availability
Sterifil Aseptic System and Holder
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Articles
Nonsterile Millex® Syringe Filters
Quality manufacturing, 100% integrity testing. Because we verify membrane integrity for 100% of Millex® filters shipped each day, you can rely on them to remove all particles from your sample.
Keywords: Chromatography, High performance liquid chromatography, Ion-exchange chromatography, Liquid chromatography mass spectrometry, Mass spectrometry
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?