• USA Home
  • Z315796 - Eppendorf® Reference® pipette, fixed volume
EMAIL THIS PAGE TO A FRIEND
Z315796 Sigma
Eppendorf® Reference® pipette, fixed volume
2 μL volume
Purchase
Properties
Related Categories Eppendorf, Eppendorf Pipettors, Eppendorf Pipettors & Tips, Labware, Liquid Handling Equipment,
material   gray control button
feature   electronic: no
  fully autoclavable
  single-channel
mfr. no.   Eppendorf®, 4900 000.028 (European)
  Eppendorf®, 4900 000.028
measuring range   2 μL volume
volume   2 μL
Description
General description
Ejector sleeve removable, for easy access to narrow vessels.
Single button control of filling, blow-out and tip ejection.
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
Reference is a registered trademark of Eppendorf AG
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?