• USA Home
  • Z316113 - Eppendorf® Mastercycler
EMAIL THIS PAGE TO A FRIEND
Z316113 Sigma
Eppendorf® Mastercycler
adapter for in-situ PCR
Purchase
Properties
mfr. no.   Eppendorf®, 5331 220.002 (European)
availability   not available in USA and Canada
Description
General description
Compact thermalcyclers with Peltier heating and cooling giving accurate, responsive temperature control.
Heated lid. Simple programming via keypad and 8-line digital display with adjustable time and temperature increments, incubation mode, touch down, link, pause and delay functions. Up to 100 programs may be stores in the cycler and individual users may stores up to ten programs on a personal card which can be used in any Eppendorf Mastercycler model for easy, reproducible transfer of work between cyclers.
Auto-restart in event of power failure.
Mastercycler and Mastercycler Gradient may be converted for in-situ PCR using a slide adapter in place of the block. Adapter holds up to 4 slides.
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?