• USA Home
  • Z317071 - Eppendorf® Combitips® Plus Biopur®
EMAIL THIS PAGE TO A FRIEND
Z317071 Sigma
Eppendorf® Combitips® Plus Biopur®
volume range 0.2 mL
Purchase
Properties
Related Categories Combitips Plus Biopur, Eppendorf, Eppendorf PCR Pipette Tips, Eppendorf Pipette Tips, Eppendorf Pipettors & Tips,
material   colorless tip
sterility   sterile
feature   ATP free
  DNA free (Bacterial (E. Coli))
  DNA free (Human)
  DNase free
  PCR inhibitor free
  RNase free
  barrier: No
  pyrogen free
packaging   pack of 100
mfr. no.   Eppendorf®, 0030 069.412 (European)
  Eppendorf®, 022496026 (USA)
availability   not available in USA and Canada
volume range   0.2 mL
Description
General description
Snaps securely into Multipette pipette. Tips are not autoclavable. Biopur tips are certified sterile and pyrogen, RNase-, DNA and ATP-free.
Legal Information
Biopur is a registered trademark of Eppendorf AG
Combitips is a registered trademark of Eppendorf AG
Eppendorf is a registered trademark of Eppendorf AG
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?