• USA Home
  • Z323195 - BRAND® wide-mouth bottles with screw cap, PTFE
EMAIL THIS PAGE TO A FRIEND
Z323195 Sigma
BRAND® wide-mouth bottles with screw cap, PTFE
capacity 10 mL
Purchase
Properties
Related Categories BRAND, BRAND Bottles, Plastic, Bottles and Caps, Fluorinated Plastic Bottles, Labware,
material   PTFE bottle
  bottle round
  bottle wide-mouth
  white bottle
feature   cap
mfr. no.   BRAND, 130508
capacity   10 mL
diam. × H   26 mm × 50 mm
neck I.D.   12 mm
Description
General description
Outstanding chemical and thermal resistance. Sturdy, strong-walled design.
Other Notes
BR130508 (India)
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?