• USA Home
  • Z323349 - BRAND® BLAUBRAND® micro burette, bang pattern, straight stopcock

EMAIL THIS PAGE TO A FRIEND
Z323349 Sigma

BRAND® BLAUBRAND® micro burette, bang pattern, straight stopcock

10 mL volume, accuracy: ±0.020 mL, borosilicate glass 3.3 (Schellbach PTFE spindle grad. in blue, stand incl.)

Purchase

Properties

Related Categories BLAUBRAND, BRAND, BRAND BLAUBRAND microburette, bang pattern, lateral stopcock, BRAND Burettes, BRAND Volumetric,
material   borosilicate glass 3.3 (Schellbach PTFE spindle grad. in blue, stand incl.)
feature   class AS
  subdivision 0.02 mL
mfr. no.   BRAND, 24599
measuring range   10 mL volume, accuracy: ±0.020 mL
burette L   900 mm
volume   10  mL

Description

General description

DURAN®. According to DIN 385, Class AS, DE-M marking. Calibrated to deliver (TD, EX + 30sec waiting time. Schellbach stripe, ring marks at major graduations. High contrast blue enamel graduations and inscriptions.

All BLAUBRAND® burettes and automatic burettes are supplied with a batch number and an accompanying batch certificate in the original packaging.

Other Notes

BR24599 (India)

Legal Information

BLAUBRAND is a registered trademark of BRAND GMBH + CO KG

BRAND is a registered trademark of BRAND GMBH + CO KG

Duran is a registered trademark of Duran Group

Price and Availability

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?