• USA Home
  • Z323462 - BRAND® SILBERBRAND burette, lateral stopcock

EMAIL THIS PAGE TO A FRIEND
Z323462 Sigma

BRAND® SILBERBRAND burette, lateral stopcock

50 mL volume, accuracy: ±0.08 mL, clear borosilicate glass 3.3 (Schellbach stripes, glass key)

Purchase

Properties

Related Categories BRAND Silberbrand burette, lateral stopcock, Brand Burettes, Labware, Volumetric, Volumetric Burettes More...
material   clear borosilicate glass 3.3 (Schellbach stripes, glass key)
grade   according to DIN 12 700-3
feature   subdivision 0.1 mL
mfr. no.   BRAND, 13508
measuring range   50 mL volume, accuracy: ±0.08 mL
L   ~800 mm
volume   50  mL

Description

General description

Class B. With Schellbach stripe. Calibrated to deliver (TS, EX). High contrast black enamel graduations and inscriptions.

PTFE valve spindle operates smoothly yet is tight, even without lubrication, so there is no grease residue on the burette. The valve spindle is replaceable. Tight seal without force, a gentle turn is all it takes to close the burette valve.

Other Notes

BR13508 (India)

Legal Information

BRAND is a registered trademark of BRAND GMBH + CO KG

Price and Availability

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?