• USA Home
  • Z327239 - BRAND® narrow-mouth bottle, LDPE
EMAIL THIS PAGE TO A FRIEND
Z327239 Sigma
BRAND® narrow-mouth bottle, LDPE
with cap: no
Synonym: ldpe bottle, polyethylene bottles, polyethylene containers
Purchase
Properties
Related Categories Bottles and Caps, Labware, Low-Density Polyethylene Bottles, Plastic Bottles
material   bottle narrow-mouth
  bottle round
  low-density polyethylene bottle
  translucent
feature   cap: no
  dropper nozzle caps available
  with cap: no
packaging   pkg of 25
mfr. no.   BRAND, 129064
diam. × H   117 mm × 264 mm
fitting   thread GL 28
Description
General description
Economical multipurpose bottle, flexible. Supplied with cap.
Other Notes
BR129064 (India)
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?