• USA Home
  • Z329142 - BRAND® magnetic stirring bar, PTFE, octagonal with pivot ring
EMAIL THIS PAGE TO A FRIEND
Z329142 Sigma
BRAND® magnetic stirring bar, PTFE, octagonal with pivot ring
L 38 mm, bar diam. 13 mm
Purchase
Properties
Related Categories 21mm to 40mm Length Octogonal Stir Bars, BRAND, BRAND Stirrers, BRAND magnetic stirring bar, PTFE, octagonal with pivot ring, Labware,
material   PTFE-coated octagonal (with pivot ring)
mfr. no.   BRAND
L   38 mm
bar diam.   13 mm
pivot ring diam.   14.5 mm
Description
General description
Angular surface creates high turbulence even at low speeds. For vessels with slightly uneven bottoms. The pivot ring reduces the contact surface and enables the stirrer to adopt the optimum position.
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?