• USA Home
  • Z329606 - BRAND® wash bottles, LDPE
EMAIL THIS PAGE TO A FRIEND
Z329606 Sigma
BRAND® wash bottles, LDPE
capacity 100 mL
Synonym: bottle wash, squeeze bottle, wash bottle
Purchase
Properties
Related Categories BRAND, BRAND Bottles, Plastic, Bottles and Caps, LDPE Wash Bottles, Labware,
material   bottle narrow-mouth
  bottle round
  low-density polyethylene bottle
  translucent bottle
feature   cap
mfr. no.   BRAND, 144138
capacity   100 mL
diam. × H   45 mm × 196 mm
fitting   neck thread GL 18
Description
General description
Wash bottles with interchangeable wash head. Flexible. Delivery tube adjustable in height. Tip can be cut back to increase flow
Other Notes
BR144138 (India)
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
recommended
Product #
Description
Add to Cart
Z329649 BRAND® wash head for wash bottles, LDPE, thread GL 18
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?