• USA Home
  • Z329940 - BRAND® narrow-mouth bottle, LDPE
EMAIL THIS PAGE TO A FRIEND
Z329940 Sigma
BRAND® narrow-mouth bottle, LDPE
capacity 30 mL
Synonym: ldpe bottle, polyethylene bottle, polyethylene container
Purchase
Properties
Related Categories BRAND, BRAND Bottles, Plastic, Bottles and Caps, Labware, Low-Density Polyethylene Bottles,
material   bottle narrow-mouth
  bottle round
  low-density polyethylene bottle
  translucent
feature   cap
  dropper nozzle caps available
packaging   pkg of 100
mfr. no.   BRAND, 129424
capacity   30 mL
diam. × H   34 mm × 66 mm
fitting   thread GL 14
Description
General description
Economical multipurpose bottle, flexible. Supplied with cap.
Other Notes
BR129424 (India)
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?