• USA Home
  • Z330396 - BRAND® standard disposable cuvettes
EMAIL THIS PAGE TO A FRIEND
Z330396 Sigma
BRAND® standard disposable cuvettes
polystyrene
Purchase
Properties
Related Categories Absorption Cells, Analytical Reagents, Analytical/Chromatography, Cuvettes, Disposable Cuvettes,
material   polystyrene
packaging   pkg of 1000 ea
mfr. no.   Brand, 759005
H × W × D   45 mm × 12.5 mm × 12.5 mm
chamber volume   2.5-4.5 mL
pathlength   10 mm
window size   10 mm × 35 mm
Description
General description
Macro or semi-micro cuvettes available in PS and PMMA. Ideal for kinetic measements. Windows are recessed to protect from scratches, arrow mark indicates optical path orientation. Plastibrand cuvettes are delivered grouped by mold cavity number to ensure lowest variation of extinction coefficient.
Useable wavelength > 340 nm
Standard deviation at 360 nm: ±0.005 extinction units
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG
Price and Availability
Just Prove it! Spectroquant Prove
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?