• USA Home
  • Z332925 - BRAND® pipette tips, bulk-packed, non-sterile
EMAIL THIS PAGE TO A FRIEND
Z332925 Sigma
BRAND® pipette tips, bulk-packed, non-sterile
volume range 50-1000 μL
  •  eCl@ss 32079201
Purchase
Properties
Related Categories BRAND, BRAND Pipettors & Tips, Brand Pipette Tips, Labware, Liquid Handling Equipment,
material   blue tip
sterility   non-sterile
feature   autoclavable
  barrier: No
  DE-M marking
packaging   case of 1000
  bag of 500
mfr. no.   BRAND, Z332925
volume range   50-1000 μL
Description
General description
Tips are produced under the latest state-of-the-art clean room conditions and automatically shrink-wrapped in bags and packaged in cardboard boxes.
For easy identification, the BRAND catalogue number, volume and lot number of the tips are found on every bag.
• High-purity PP (free from DiHEMDA and oleamide)
• Weight reduction of the 200 μL and 1000 μL tips to reduce the weight of plastic waste
• Graduations for a quick volume check
• Autoclavable at 121°C (2 bar) acc. DIN EN 285
• Environmentally friendly packaging
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?