• USA Home
  • Z333069 - BRAND® pipette tips, Tip-Rack, non-sterile
EMAIL THIS PAGE TO A FRIEND
Z333069 Sigma
BRAND® pipette tips, Tip-Rack, non-sterile
volume range 5-300 μL
Purchase
Properties
Related Categories BRAND, BRAND Pipettors & Tips, Brand Pipette Tips, Labware, Liquid Handling Equipment,
material   colorless tip
  green racks
description   Two individual refill units (racks) are packed into a space-saving carton
sterility   non-sterile
feature   barrier: no
  conformity certified.
packaging   pack of 960
mfr. no.   BRAND, 702353
volume range   5-300 μL
Description
General description
Refill units for Tip-Box n, Tip-Box SL or Tip-Box PC. Two filled tip-trays (racks) are packed into a space-saving carton. Up to 300 μL: racks with 96 tips and 1000 μL: racks with 60 or 100 tips each, 1250 μL: racks with 100 tips.
• High-purity PP (free from DiHEMDA and oleamide)
• Weight reduction of the 200 μL and 1000 μL tips to reduce the weight of plastic waste
• Graduations for a quick volume check
• Autoclavable at 121°C / 250°F (2 bar), acc. DIN EN 285
• Environmentally friendly packaging
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?