• USA Home
  • Z333093 - BRAND® Tip-Box N, filled with pipette tips
EMAIL THIS PAGE TO A FRIEND
Z333093 Sigma
BRAND® Tip-Box N, filled with pipette tips
volume range 5-20 μL
Purchase
Properties
Related Categories BRAND, BRAND Pipettors & Tips, Brand Pipette Tips, Labware, Liquid Handling Equipment,
material   polypropylene
  colorless tip
  gray rack
sterility   non-sterile
feature   barrier: No
packaging   case of 480
mfr. no.   BRAND, 702411
volume range   5-20 μL
Description
General description
With push-on lid, stackable.
• Two differnet heights: Low form (up to 300 μL): rack with 96 tips, High form (1000 μL or 1250 μL): rack with 100 tips
• High-quality PP (fre from DiHRMDA and oleamide)
• Weight reduction of the 200 μL and 1000 μL tips to reduce the weight of plastic waste
• Graduations for quick volume check
• Autoclavable at 121°C/250°F (2 bar), acco DIN EN 285
• Environmentally friendly packaging
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?