• USA Home
  • Z333263 - BRAND® filter tips, bulk-packed, non-sterile
EMAIL THIS PAGE TO A FRIEND
Z333263 Sigma
BRAND® filter tips, bulk-packed, non-sterile
volume range 5-100 μL
Purchase
Properties
Related Categories Aerosol Barrier Pipette Tips, BRAND, BRAND Pipettors & Tips, Brand Pipette Tips, Filter Tips,
material   colorless tip
sterility   non-sterile
feature   autoclavable at 121 °C
  barrier
packaging   bag of 960
  bag of 960
mfr. no.   BRAND, 702104
volume range   5-100 μL
Description
General description
Filter tips are especially suited to working with PCR techniques, and fully meet the requirements for microbiology and radio-isotope work. The integrated hydrophobic polypropylene-filter serves as a barrier against aerosols formed during pipetting.
These non-self sealing filter tips protect the pipette shafts from contamination, and thus lessen the risk of cross-contamination and faulty measuring results.
• High-purity PP (free from DiHEMDA and oleamide)
• Weight reduction of the 20 μL tips to reduce the weight of plastic waste
• Graduations for a quick volume check
• Autoclavable at 121°C/250°F (2 bar) acc. DIN EN 285
• Environmentally friendly packaging
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?