• USA Home
  • Z333336 - BRAND® BIO-CERT® filter tips, Tip-Rack S, sterile
EMAIL THIS PAGE TO A FRIEND
Z333336 Sigma
BRAND® BIO-CERT® filter tips, Tip-Rack S, sterile
volume range 5-100 μL
Purchase
Properties
Related Categories BRAND, BRAND Pipettors & Tips, Brand Pipette Tips, Filter Tips, Filtered,
material   colorless tip
sterility   (autoclavable at 121°C)
  sterile
  sterile
feature   ATP free
  DNA free
  RNase free
  barrier
  endotoxin free
packaging   rack of 96
mfr. no.   BRAND, 702144
volume range   5-100 μL
Description
General description
1 loaded BIO-CERT refill unit for the autoclaved Tip-Box N Tip-Box SL or Tip--Box PC. Protected in an environmentally compatible packaging from recyclable PET. (Tip-Rack S is only available for volume ranges up to 300 μL)
• High-purity PP (free from DiHEMDA and oleamide)
• Weight reduction of the 20 μL tips to reduce the weight of plastic waste
• Graduations for a quick volume check
• BIO-CERT: sterile, free of DNA, DNase, RNase, endotoxins and ATP
• Autoclavable at 121 °C / 250 °F (2 bar), acc. DIN EN 285
• Environmentally friendly packaging
Legal Information
BIO-CERT is a registered trademark of BRAND GMBH + CO KG
BRAND is a registered trademark of BRAND GMBH + CO KG
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?