• USA Home
  • Z338826 - Maxi horizontal electrophoresis units
EMAIL THIS PAGE TO A FRIEND
Z338826 Sigma
Maxi horizontal electrophoresis units
uncooled unit
Purchase
Description
General description
Designed for both analytical and preparative separations of nucleic acids with buffer circulation ports to prevent buffer depletion. Four comb positions permit the screening of up to 160 samples.
Packaging
Includes base unit, safety lid, one standard casting tray for 20 × 20 cm gels, and two combs, 1.0 mm × 16 sample wells
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?