• USA Home
  • Z338850 - Maxi-Plus horizontal electrophoresis units
EMAIL THIS PAGE TO A FRIEND
Z338850 Sigma
Maxi-Plus horizontal electrophoresis units
cooled unit
Purchase
Properties
Related Categories Electrophoresis Supplies, Horizontal Gel Electrophoresis Supplies, Labware, Units and Components for Horizontal Gel Electrophoresis
feature   with hose connectors for circulating coolant
gel W   25-30 cm
Description
General description
This electrophoresis unit has been specifically designed to allow quick and convenient screening of PCR products. When using a 96 well PCR machine, all samples can be screened on a single gel. Loading is simple using a standard multichannel pipette.
The unit is constructed of 12 mm thick acrylic to prevent warping and is supplied with a UV transparent tray for easy visualization. For varying sample numbers a range of alternative combs can be used. As there are 12 comb positions, up to 624 samples can be separated on a single gel.
Includes base unit, safety lid, one standard casting tray for 25 x 30 cm gels, and two combs, 1.0 mm x 26 sample wells.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?