• USA Home
  • Z338885 - Comb for mini horizontal electrophoresis unit
EMAIL THIS PAGE TO A FRIEND
Z338885 Sigma
Comb for mini horizontal electrophoresis unit
size 1.0 mm × 12 wells
Purchase
Properties
Related Categories Comb for mini horizontal electrophoresis unit, Combs for Horizontal Gel Electrophoresis Units, Electrophoresis Supplies, Horizontal Gel Electrophoresis Supplies, Labware More...
feature   well spacing compatible with multichannel pipettes
size   1.0 mm × 12 wells
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?