• USA Home
  • Z339547 - Maxi-Plus horizontal electrophoresis unit accessory
EMAIL THIS PAGE TO A FRIEND
Z339547 Sigma
Maxi-Plus horizontal electrophoresis unit accessory
Colored gel loading strips
Purchase
Properties
packaging   pk of 6 (2 colors)
Description
Compatibility
Provide a colored background behind sample loading area making sample wells more visible. PAK = 6, 2 colors.
General description
Multichannel pipette compatible
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?