• USA Home
  • Z339571 - Mini Vertical Electrophoresis Unit
EMAIL THIS PAGE TO A FRIEND
Z339571 Sigma
Mini Vertical Electrophoresis Unit
Set with Cooling block
Purchase
Description
Components
Included are 1.0 mm × 10 sample combs and matching spacers (two sets).
General description
• High quality acrylic construction with replaceable platinum electrode wires
• Safety lid has integral shrouded power leads with corrosion-resistant gold-plated terminals
• All dual-gel models for higher throughput
• Casting base (included) uses cams and silicone rubber pads to hold fully assembled running module and two gel sandwiches in perfect alignment. After gel sets, remove entire assembly and place in tank.
• Running module holds gels with easy-to-use plate-clamping bars and screws
• Wide range of matching combs and spacers
Each unit comes with tank and lid, power leads and connectors, dual-gel running module, casting base, four sets of glass plates, one acrylic dummy plate (for running in single-gel mode), two sample combs, two spacer sets, and two spacer aligners. All components are available separately
Plates 105 mm x 100 mm, usable gel size 80 mm x 85 mm
The cooling block version has hose connectors for tap water or coolant from a circulating water bath. Cooled gels can be run faster than conventional gels, minimizing heat induced lane distortions, and are ideal for native protein separations as well as SDS-Protein and DNA applications requiring temperature control.
Price and Availability
Detect Your Unseen Protein
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?