• USA Home
  • Z339628 - Mini Vertical Electrophoresis Unit accessories
EMAIL THIS PAGE TO A FRIEND
Z339628 Sigma
Mini Vertical Electrophoresis Unit accessories
Acrylic Dummy Plate
Purchase
Description
General description
Suitable for TV100 and old mini-vertical units (Z339563 and Z339571)
Other Notes
For running mini-vertical unit in single-gel mode
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?