• USA Home
  • Z340588 - BRAND® BISTABIL® ground glass stopper, conical joint
EMAIL THIS PAGE TO A FRIEND
Z340588 Sigma
BRAND® BISTABIL® ground glass stopper, conical joint
Joint: ST/NS 19/26
Purchase
Properties
Related Categories Bistabil ground glass stopper, Glass Stoppers, Labware, Lenz, Manufacturer Browser,
Joint   Joint: ST/NS19/26
mfr. no.   BRAND, 144625
Description
General description
Conical ground joint stopper made from Duran®. Hollow with drip tip and hexagonal grip. Meets DIN 12 252.
Other Notes
BR144625 (India)
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG
Bistabil is a registered trademark of Lenz GmbH
Duran is a registered trademark of Duran Group
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?