• USA Home
  • Z350842 - Transfer pipette, polyethylene

EMAIL THIS PAGE TO A FRIEND
Z350842 Sigma

Transfer pipette, polyethylene

graduated to 2 mL, blood bank, bulb draw 1.8 mL, sterile

Purchase

Properties

Related Categories Labware, Liquid Handling Equipment, Pipettes, Pipettes, Pipettors and Tips, Transfer Pipettes More...
material   polyethylene
sterility   sterile
mfr. no.   Sigma-Aldrich
L   156 mm
bulb draw   1.8 mL
stem diam.   7 mm

Description

General description

Disposable transfer pipettes are a great way to transfer liquids without the worry of cross-contamination. Available in 19 styles, our extensive range is sure to have one that fits your needs.

Available as non-sterile, sterile, or sterile and wrapped in packs of 10.

• Unified design with built-in pipette bulb
• Safe, convenient use and disposal
• Reduces loss of cells and proteins that bind to glass
• Heat sealable to create liquid-tight storage
• Non-autoclavable

Packaging

Packaged in bags of 20

Price and Availability

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?