• USA Home
  • Z351059 - Transfer pipette, polyethylene
EMAIL THIS PAGE TO A FRIEND
Z351059 Sigma
Transfer pipette, polyethylene
narrow two-step stem
Purchase
Description
General description
Disposable transfer pipettes are a great way to transfer liquids without the worry of cross-contamination. Available in 19 styles, our extensive range is sure to have one that fits your needs.
Available as non-sterile, sterile, or sterile and wrapped in packs of 10.
• Unified design with built-in pipette bulb
• Safe, convenient use and disposal
• Reduces loss of cells and proteins that bind to glass
• Heat sealable to create liquid-tight storage
• Non-autoclavable
Packaging
Same as Z351032, but sterile, packaged in bags of 20
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
comparable product
Product #
Description
Add to Cart
Z351040 Transfer pipette, polyethylene, narrow two-step stem, bulb draw 3.0 mL, sterile
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?