• USA Home
  • Z351083 - Hanna hand-held thermometer
EMAIL THIS PAGE TO A FRIEND
Z351083 Sigma
Hanna hand-held thermometer
Model HI 8753
Purchase
Properties
Related Categories Electronic Meters, Hanna, Hanna Hand-Held Thermometer, Hanna Temperature measurement, Labware,
feature   display 4 digit
mfr. no.   Hanna
measuring range   −40-+150 °C temperature, accuracy: 0.5% full scale, resolution: 0.1 °C
size   180 mm (7.1 in.) × 83 mm (3.3 in.) × 40 mm (1.6 in.)
weight   280 gwith probe
Description
General description
• NTC thermometer embedded in a stainless steel penetration probe allows wide temperature range measurement without changing probes
• reads °C or °F
This model is supplied complete with HI765BL temperature probe with 1 m (3.3 ft) cable, 9 V battery and instruction manual.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?