Z351091 Sigma
Hanna Checker® pH meter
Checker 1
DOWNLOAD MSDS (PDF)
Properties
Related Categories Hand Held, Hanna, Hanna Checker pH meter, Hanna pH Meters, Hanna pH Meters and Accessories,
feature   calibration two point manual with trimmers
  temp. range 0 ‑ 50 °C
measuring range   pH, accuracy: 0.2 pH unit, (at 20 deg C)
  0-14 pH, resolution: 0.01
Description
General description
Features:
• Lightweight, pocket-sized
• Supplied complete with a replaceable single-junction combination electrode (see below)
• 2 × 1.4 V button cells give 3000 hrs operation
With replaceable screw-type, combination electrode
Legal Information
Checker is a registered trademark of Hanna Instruments
Price and Availability
Customers Also Viewed
Hanna pH Meter model HI 9024
0-14 pH, accuracy: 0.01, resolution: 0.01, 0-100 °C temperature, accuracy: 0.5 °C
Hanna pH meter model HI 9025C
0-14 pH, accuracy: 0.01, battery power 4 × 1.5 V AA, 400 hr life
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?