• USA Home
  • Z351105 - Hanna pH meter model HI 9025C
EMAIL THIS PAGE TO A FRIEND
Z351105 Sigma
Hanna pH meter model HI 9025C
0-14 pH, accuracy: 0.01, battery power 4 × 1.5 V AA, 400 hr life
Purchase
Properties
Related Categories Hand Held, Hanna, Hanna pH Meters, Hanna pH Meters and Accessories, Hanna pH model HI 9025C,
mfr. no.   Hanna
measuring range   −1999-1999 mV ORP, accuracy: 1
  −399.9-399.9 mV ISE, accuracy: 0.1
  0-14 pH, accuracy: 0.01
Description
General description
Heavy duty, waterproof portable pH/mv/° C Meter. BNC connector.
• Temperature: manual or automatic
• probe included
• dual display
• Calibration: automatic one- or two-point
• display prompts for buffers
• preprogrammed with five NBS and European standard buffers
• READY indicator for accurate calibration
• Completely waterproof, totally immersible meter with rugged ABS case.
• Excellent for field
• Memory
• Automatic switch-off after 10 min (can be overridden)
• Low battery indicator
• 500 hr use with 4 AAA batteries
· Supplied complete with double-junction combination electrode, carrying case and buffers pH 4 and 7.
Help in selecting the correct electrode can be viewed at Electrode Selection Guide
Hints on Electrode Maintenance
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?