• USA Home
  • Z351393 - Eppendorf® Repeater pipette tips
EMAIL THIS PAGE TO A FRIEND
Z351393 Sigma
Eppendorf® Repeater pipette tips
Combitip combination set
Purchase
Properties
sterility   sterile; autoclavable (ultra-micro tip only)
mfr. no.   Eppendorf®
  Eppendorf®
Description
Features and Benefits
Includes 20 each of the 0.5, 1.25, 2.5, 5.0, and 12.5 mL sizes.
General description
The Combitip′s integral reservoir lets you pipet up to 48 times without refilling. Liquids contact only the reusable, disposable Combitip, eliminating cleaning and contamination of the pipet itself. Choice of Combitips gives volume selection from 1 μl to 5 mL.
The Biopur tips are sterile and certified free of DNA, RNASE, ATP, and pyrogen.
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
Repeater is a trademark of Brinkmann Instruments, Inc.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?