• USA Home
  • Z351431 - Eppendorf® Biomaster® 4830 PCR pipette
EMAIL THIS PAGE TO A FRIEND
Z351431 Sigma
Eppendorf® Biomaster® 4830 PCR pipette
Model 4830
Purchase
Properties
Related Categories Eppendorf, Eppendorf PCR, Eppendorf PCR Pipettor & tips, Eppendorf Pipettors, Labware,
feature   electronic: no
  single-channel
mfr. no.   Eppendorf®, 022440501 (USA)
  Eppendorf®, 4830 000.017 (European)
  Eppendorf®, 4830 000.017
measuring range   1-20 μL volume
volume   20 μL
Description
General description
Volume is adjustable from 1-20 μl in 0.02 μl increments. Volume setting automatically locks in, eliminating accidental changes. UV resistant and autoclavable with a built-in tip ejector. Integrated disposable tip and piston assembly reduces sample contamination by eliminating aerosols and sample carryover.
Legal Information
Biomaster is a registered trademark of Eppendorf AG
Eppendorf is a registered trademark of Eppendorf AG
Price and Availability
Your Key to Success
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?