• USA Home
  • Z351482 - Eppendorf® Pipette tips Ultra-micro
EMAIL THIS PAGE TO A FRIEND
Z351482 Sigma
Eppendorf® Pipette tips Ultra-micro
Biopur, volume range 20 μL
Purchase
Properties
Related Categories Eppendorf Pipette Tips, Labware, Liquid Handling Equipment, Pipette Tips, Pipettes, Pipettors and Tips More...
material   colorless tip
sterility   sterile
feature   ATP free
  DNA free (Bacterial (E. Coli))
  DNA free (Human)
  DNase free
  PCR inhibitor free
  RNase free
  barrier: no
  pyrogen free
packaging   rack of 96
mfr. no.   Eppendorf®, 0030 065.506 (European)
  Eppendorf®, 022491067 (USA)
volume range   20 μL
Description
Features and Benefits
Certified DNA-, RNase-, ATP-, and pyrogen-free. Automated production and packaging prevent contamination from microorganisms and human contact. Molded from dye-free resins.
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?