• USA Home
  • Z351598 - Eppendorf® pipette tips
EMAIL THIS PAGE TO A FRIEND
Z351598 Sigma
Eppendorf® pipette tips
Eppendorf Envirotips, volume range 200 μL
Purchase
Properties
Related Categories Eppendorf Pipette Tips, Labware, Liquid Handling Equipment, Pipette Tips, Pipettes, Pipettors and Tips More...
material   colorless tip
sterility   non-sterile
feature   barrier: No
packaging   rack of 96
mfr. no.   Eppendorf®, 022490427 (USA)
volume range   200 μL
Description
General description
Dye-free tips racked in 8 × 12 in. compact plastic trays held in biodegradable, recyclable cardboard packaging. The individual package consists of a double-sided box with a tray full of tips on each side. Tips can be used directly from the package or from an Envirobox.
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?