• USA Home
  • Z351695 - Pipette tips, disposable, polypropylene, capacity 1000 μL
EMAIL THIS PAGE TO A FRIEND
Z351695 Sigma
Pipette tips, disposable, polypropylene, capacity 1000 μL
Eppendorf Enviroset
Purchase
Properties
packaging   package of 192 tips
mfr. no.   Eppendorf®
  Eppendorf®
Description
General description
Dye-free. Racked in 8x12 compact plastic trays held in biodegradable, recyclablecardboard packaging. Individual packages consist of a double-sided box with a tray full of tips on each side. Tips can be used directly from the package or an Envirobox.
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
Price and Availability
Standard component
Description
Product #
Add to Cart
192 tips 2 packs SDS    
Envirobox with one Envirotip tray 1 SDS    
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?