• USA Home
  • Z351741 - Eppendorf® pipette tips
EMAIL THIS PAGE TO A FRIEND
Z351741 Sigma
Eppendorf® pipette tips
Eppendorf Enviroset, capacity 2500 μL
Purchase
Properties
Related Categories Eppendorf Pipette Tips, Labware, Liquid Handling Equipment, Pipette Tips, Pipettes, Pipettors and Tips More...
mfr. no.   Eppendorf®
  Eppendorf®
capacity   2500 μL
Description
Features and Benefits
Envirobox has a sliding cover which allows row-by-row use (eight tips at a time) without exposing the entire tray.
Packaging
Package of one Envirobox and 336 tips
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
comparable product
Product #
Description
Add to Cart
Z640379 Eppendorf® epT.I.P.S. set, volume range 500-2500 μL
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?