• USA Home
  • Z351784 - Eppendorf® Maxitips for Maxipettor®
EMAIL THIS PAGE TO A FRIEND
Z351784 Sigma
Eppendorf® Maxitips for Maxipettor®
Maxitip P, non-sterile
Purchase
Properties
Related Categories Eppendorf, Eppendorf Pipette Tips, Eppendorf Pipettors & Tips, Eppendorf Tips, Labware,
packaging   pkg of 100 assemblies
mfr. no.   Eppendorf®, 0030 048.130 (European)
  Eppendorf®, 022291050 (USA)
volume   1-10 mL
Description
General description
These tips allow the Maxipettor to function like an air-displacement serological pipet with integral pipetting aid. Reusable barrel/piston assembly attaches to the Maxipettor, and 170 mm Maxitip S or G attaches to the barrel.
Specially designed valve can be added to the assembly to allow dripless pipetting of liquids with high vapor pressure.
Product features of Maxitips:
• Maxitip P for aspirating 1 mL to 10 mL from beakers using a positive-displacement technique
• Maxitip S for aspirating 2.5 mL to 10 mL from tall, narrow-neck vessels using an air-displacement technique
• No carryover as Maxitips are exchanged
• Valve for Maxitip ensures drip-free dispensing of liquids with a high vapor pressure
• Maxitip G is graduated for dispensing accurate volumes of non-aqueous liquids
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
Maxipettor is a registered trademark of Eppendorf AG
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?