• USA Home
  • Z351857 - Sigma-Aldrich® nucleic acid sequencing units
EMAIL THIS PAGE TO A FRIEND
Z351857 Sigma
Sigma-Aldrich® nucleic acid sequencing units
Fixed height, single gel, gel W × H 20 cm × 52 cm
Purchase
Properties
Related Categories Electrophoresis Supplies, Labware, Units /Gel Box, Vertical Gel Electrophoresis Units
availability   not available in EU
gel W × H   20 cm × 52 cm
Description
Features and Benefits
Special features of these units are:
• safety interlock lids
• detachable upper and lower buffer reservoirs
• upper and lower reservoirs have drains with tubing adapters for easy and safe disposal of buffer solutions
• leveling base with bubble level
Each unit comes complete with clamps, sealing tape, power leads, and glass plates.
Supplied with two sets of glass plates, bar clamp, and a built-in epoxy-coated aluminum cooling plate.
Legal Information
Sigma-Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?