• USA Home
  • Z351865 - Sigma-Aldrich® nucleic acid sequencing units
EMAIL THIS PAGE TO A FRIEND
Z351865 Sigma
Sigma-Aldrich® nucleic acid sequencing units
Adjustable Height, Single Gel, gel W × H 20 cm × 22-82 cm
Purchase
Properties
Related Categories Electrophoresis Supplies, Labware, Units /Gel Box, Vertical Gel Electrophoresis Units
availability   not available in EU
gel W × H   20 cm × 22-82 cm
Description
Features and Benefits
Special features of these units are:
• safety interlock lids
• detachable upper and lower buffer reservoirs
• upper and lower reservoirs have drains with tubing adapters for easy and safe disposal of buffer solutions
• leveling base with bubble level
Each unit comes complete with clamps, sealing tape, power leads, and glass plates.
The upper buffer reservoir slides on three 1-inch Delrin rods and is secured by stainless steel thumbscrews at any height from 22 to 82 cm. Supplied with 1set each of 42cm and 82cm glass plates. Cooling plate not included.
Legal Information
Sigma-Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?